Orders LinkContactUsLinkComponent
Dunke
Dunke
10 L, UN approval: UN 3H1/Y/1.9, 195×225×320 mm, GL 51
Katalog # KART1782
Leverandør:   KARTELL
Dunke
Katalog # KART1782
Leverandør:  KARTELL
Dunke
Dunke
10 L, UN approval: UN 3H1/Y/1.9, 195×225×320 mm, GL 51
Price
4.240,00 kr
Log ind for tilgængelighed
1 * 24 Items
Product Details & Documents
HDPE, translucent with black screw cap.
  • With comfortable handle and secure seal cap following DIN recommendations
  • Conform to UN regulation on transportation of dangerous liquids
  • Suitable for foodstuff
Ideal for storing and transporting liquids.
Specifikationer
  • Gevind:
    GL 51
  • Kapacitet:
    10 L
  • Beskrivelse:
    UN approval: UN 3H1/Y/1.9
  • B×D×H:
    195×225×320 mm