Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Industriøser, plastilab®

Industriøser, plastilab®

1000 ml, 350 mm
Katalog # KART1066
Leverandør:   KARTELL
Industriøser, plastilab®
Katalog # KART1066
Leverandør:  KARTELL
Industriøser, plastilab®
Industriøser, plastilab®

1000 ml, 350 mm

Price
1.330,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents

These scoops made from HDPE are designed for heavy industrial use.

  • Round shaped design facilitates complete transfer of sampling material
  • Suitable for foodstuff
Specifikationer
  • Kapacitet:
    1000 ml
  • Længde:
    350 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.