Funnels, powder
80 mm
Catalog # WITG5.790.004
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Tragt udv. Ø:80 mm