Orders LinkContactUsLinkComponent
Condensers, Graham
160 mm, 19/26 NS, - NS
Katalog # WITG1.732.160
Leverandør:   witeg Labortechnik
Condensers, Graham
Katalog # WITG1.732.160
Leverandør:  witeg Labortechnik
Condensers, Graham
Condensers, Graham
160 mm, 19/26 NS, - NS
Price
886,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Coil type, Graham pattern, suitable for either reflux or distillation work. Water connection with glass olives.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kondenserlængde:
    160 mm
  • Kappestr.:
    - NS