Orders LinkContactUsLinkComponent
Distilling heads
Distilling heads
24/29 NS, 24/29 NS
Katalog # WITG1.010.005
Leverandør:   witeg Labortechnik
Distilling heads
Katalog # WITG1.010.005
Leverandør:  witeg Labortechnik
Distilling heads
Distilling heads
24/29 NS, 24/29 NS
Price
412,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Specifikationer
  • Konus til kondenser:
    24/29 NS
  • Konus til kolbe:
    24/29 NS