Distilling heads
14/23 NS, 14/23 NS
Katalog # WITG1.010.001
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Specifikationer
- Konus til kondenser:14/23 NS
- Konus til kolbe:14/23 NS