Vippemåler
50 ml
Catalog # BRND43078
Supplier: Brand
Jump to:
Product Details & Documents
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
- Calibrated to deliver (TD, Ex)
- Excellent chemical resistance
- Excellent good temperature resistance
Automatic dispenser particularly suited for dispensing suspensions.
Ordering information: Reservoir bottles available on request.
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
Specifications
- Kapacitet:50 ml
- Seq:05
Accessories
Product Information
- Catalog #BeskrivelseTil NSAvailabilityPrice
Product Information
- Catalog #BeskrivelseKapacitetNSAvailabilityPrice
- BRND126963Reservoirflasker til flaskedispensere, borosilikatglas 4,1, NS 29/321000 ml29/32Log in for availability1 * 1 Items - 188,00 kr
Product Information
- Catalog #Til NSAvailabilityPrice