Biopsy processing/embedding cassettes, Swingsette®, M516
blå
Katalog # 720-1652
Leverandør: Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M516
Katalog # 720-1652
Leverandør: Simport Scientific
Gå til:
Product Details & Documents
Acetal polymer.
- Integral hinged lid, with large tab for easier opening
- Disposable plastic cassettes are highly resistant to histological solvents
- Small, square openings ensure good fluid exchange without having to use foam pads or biopsy bags
- Front face inclined 45° enabling use with certain automatic labelling systems
: Each case contains three boxes of 500 cassettes.
Specifikationer
- Farve:blå