Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Swingsette®, M516
Biopsy processing/embedding cassettes, Swingsette®, M516
lilla
Katalog # 720-1642
Leverandør:   Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M516
Katalog # 720-1642
Leverandør:  Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M516
Biopsy processing/embedding cassettes, Swingsette®, M516
lilla
Price
2.710,00 kr
Log ind for tilgængelighed
1 * 1.500 Items
Product Details & Documents
Acetal polymer.
  • Integral hinged lid, with large tab for easier opening
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Small, square openings ensure good fluid exchange without having to use foam pads or biopsy bags
  • Front face inclined 45° enabling use with certain automatic labelling systems
Packaging: Each case contains three boxes of 500 cassettes.
Specifikationer
  • Farve:
    lilla