Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat ≥99%

Bariumnitrat ≥99%

Glass bottle for solids, 500 g
Catalog # A11305.36
Oxidizing
Irritant
Bariumnitrat ≥99%
Catalog # A11305.36
CAS Number:  10022-31-8
Bariumnitrat ≥99%
Bariumnitrat ≥99%

Glass bottle for solids, 500 g

CAS Number:  10022-31-8
Price
413,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Melting Point: 592° dec. Merck Reference: 14,983 MDL No.: MFCD00003442

  • CAS Number:10022-31-8
  • Formula:Ba(NO₃)₂
  • MDL Number:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:14,983
  • Melting Pt:592 °C (dec.)
  • Density:3,24
  • Storage Temperature:Stue temperatur
  • EINECS:233-020-5
  • UN:1446
  • ADR:5.1,II
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.