Vippemåler
25 ml
Katalog # BRND43070
Leverandør: Brand
Gå til:
Product Details & Documents
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
- Calibrated to deliver (TD, Ex)
- Excellent chemical resistance
- Excellent good temperature resistance
Automatic dispenser particularly suited for dispensing suspensions.
Bestillingsoplysninger: Reservoir bottles available on request.
Documents
Brand Volumetric Measurement in the Laboratory
Produktlitteratur
Specifikationer
- Kapacitet:25 ml
- Seq:04
Accessories
Produktinformation
- Katalog #Til NSTilgængelighedPrice
- Joint clamp
Produktinformation
- Katalog #KapacitetNSTilgængelighedPrice
- Reservoirflasker til flaskedispensere, borosilikatglas 4,1, NS 29/32Reservoirflasker til flaskedispensere, borosilikatglas 4,1, NS 29/32BRND1269631000 ml29/32Log ind for tilgængelighed1 * 1 Items - 188,00 kr
Produktinformation
- Katalog #Til NSTilgængelighedPrice