Cover glasses, square
24×24 mm, 0,13 - 0,16 mm, #1, Borosilikatglas
Katalog # HECH40990121
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Borosilicate glass coverslips for histology and microscopy.
Specifikationer
- Antal:#1
- L×B:24×24 mm
- Materiale:Borosilikatglas
- Tykkelse:0,13 - 0,16 mm