Sulfonation flasks, with three necks
350 ml, 29/32 NS, 14/23 NS
Katalog # WITG0.851.024
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifikationer
- Kapacitet:350 ml
- Seq:03
- Sidehalse:14/23 NS
- Centreret hals:29/32 NS