PCR racks
96-brønds PCR-stativ, blå
Katalog # RATI6114102
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
Racks made of PP and are characterised by their very thin but uniform walls.
- 8×12 positions, micro test plate format
- Autoclavable
- Available in different colors for the convenient identification of different types of samples
- Transparent lids available in two different heights (Lids must be ordered seperately)
Work and storage rack for 0,2 ml and 0,5 ml PCR tubes, PCR strips, and PCR segments.
Specifikationer
- Farve:blå
- Beskrivelse:96-brønds PCR-stativ
- Seq:02
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Låg, transparent, højde 14 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Låg, transparent, højde 28 mm