Orders LinkContactUsLinkComponent
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Disc sensor (alarm at empty state)
Katalog # 590-0734
Leverandør:   S.C.A.T.
Accessories for SCAT Level Controls
Katalog # 590-0734
Leverandør:  S.C.A.T.
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Disc sensor (alarm at empty state)
Price
2.300,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
  • Switching cables
  • Disk sensors
  • Signal cables
  • Power supply units
Specifikationer
  • Beskrivelse:
    Disc sensor (alarm at empty state)
  • Seq:
    12