Orders LinkContactUsLinkComponent
Flange clamps
Flange clamps
Ball and flat flanges KF/PF 25
Katalog # LENZ06248525
Leverandør:   Lenz Laborglas
Flange clamps
Katalog # LENZ06248525
Leverandør:  Lenz Laborglas
Flange clamps
Flange clamps
Ball and flat flanges KF/PF 25
Price
877,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Plastic, black.
  • Easy to assemble and disassemble
  • Tough construction, good temperature resistance
  • Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Specifikationer
  • Til:
    Ball and flat flanges KF/PF 25
  • Seq:
    02