Flange clamps
Ball and flat flanges KF/PF 25
Katalog # LENZ06248525
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Plastic, black.
- Easy to assemble and disassemble
- Tough construction, good temperature resistance
- Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Specifikationer
- Til:Ball and flat flanges KF/PF 25
- Seq:02