Thread adapters for pumps
Intrenal thread adapter, PP, white, 63 mm - 50,8 mm BSP
Katalog # BURK0600-2006
Leverandør: BURKLE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Intrenal thread adapter, PP, white, 63 mm - 50,8 mm BSP
- Seq:10