Accessories
APH 530 Connection cable compatible with APH 510 PRT110 report printer, length: 10 m, Ø 10 mm, PG 8535, PG 8536, PG 8583, PG 8583 CD, PG 8593
Katalog # 141-0491
Leverandør: MIELE
Gå til:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifikationer
- Beskrivelse:APH 530 Connection cable compatible with APH 510 PRT110 report printer, length: 10 m, Ø 10 mm
- Seq:02
- Til:PG 8535, PG 8536, PG 8583, PG 8583 CD, PG 8593