Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
ICP-MS tuning solutions

ICP-MS tuning solutions

ICP-MS stock tuning solution, 5 elements in diluted nitric acid, Li, Y, Ce, Tl, Co, 10 mg/L, 2% HNO₃, 100 mL
Katalog # HEWL5188-6564
Leverandør:   AGILENT HEWLETT PACKARD
ICP-MS tuning solutions
Katalog # HEWL5188-6564
CAS nummer:  
ICP-MS tuning solutions
ICP-MS tuning solutions

ICP-MS stock tuning solution, 5 elements in diluted nitric acid, Li, Y, Ce, Tl, Co, 10 mg/L, 2% HNO₃, 100 mL

CAS nummer:  
Price
1.710,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

PA Tuning solution set includes Tuning 1 and Tuning 2 solutions.

Tuning 1: 20 ppm each of Zn, Be, Cd, As; 10 ppm each of Ni, Pb, Mg; 5 ppm each of Tl, Na, Al, U, Cu, Th, Ba, Co, Sr, V, Cr, Mn, 6Li, Sc, In, Lu, Bi; 2,5 ppm each of Y, Yb; matrix 2 to 5% HNO₃
Tuning 2: 10 ppm each of Mo, Sb, Sn, Ge, Ru, Pd; 5 ppm of Ti, Ir; matrix 10% HCl and 1% HNO₃ with trace amounts of HF.

Documents

Specifikationer

Specifikationer

  • Koncentration:
    10 mg/L
  • Matrice:
    2% HNO₃
  • Pakn:
    100 mL
  • Beskrivelse:
    ICP-MS stock tuning solution, 5 elements in diluted nitric acid, Li, Y, Ce, Tl, Co

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.