Orders LinkContactUsLinkComponent
Headspace screw top vials
Headspace screw top vials
10,00 ml, 23×46 mm, klar, No
Catalog # HEWL5188-5392
Headspace screw top vials
Catalog # HEWL5188-5392
Headspace screw top vials
Headspace screw top vials
10,00 ml, 23×46 mm, klar, No
Price
612,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
The CombiPAL precision-threaded headspace screw top vials are ideal for applications in the environmental, food and beverage, industrial hygiene, drug analysis, and clinical chemistry industries.
Documents
Specifications
  • Kasket inkluderet:
    No
  • Diameter×H:
    23×46 mm
  • Kapacitet:
    10,00 ml
  • Farve:
    klar
  • Seq:
    01