Orders LinkContactUsLinkComponent
Caps for tubes
Caps for tubes
Cap for 15 mm tubes, ufarvet
Katalog # DELT307202
Leverandør:   DELTALAB
Caps for tubes
Katalog # DELT307202
Leverandør:  DELTALAB
Caps for tubes
Cap for 15 mm tubes, ufarvet
Price
767,00 kr
Log ind for tilgængelighed
1 * 2.500 Items
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Specifikationer
  • Farve:
    ufarvet
  • Beskrivelse:
    Cap for 15 mm tubes
  • Seq:
    12