HPLC columns, ZORBAX® Eclipse Plus
Analytisk, C18, 95 Å, 1,8 µm, 4,60 mm, 50 mm
Katalog # 554-1420
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Eclipse Plus columns are recommended for initial method development from pH 2 to 9 and provide high resolution and excellent peak shape of all types of compounds.
- High level of performance (peak shape, efficiency, resolution and lifetime) with all sample types: acids, bases and neutrals
- Superior reproducibility with more rigorous QA/QC testing
- Improved, patented silica manufacturing with start-to-finish product control
Specifikationer
- Kolonne form:Analytisk
- Ø indv.:4,60 mm
- Partikelstørrelse:1,8 µm
- Porestørrelse:95 Å
- Længde:50 mm
- Seq:07
- Fase:C18
- Cat. No.:554-1420
- Beskrivelse:C18
- Endcapped:Dobbelt
- USP code:L1
- Temperature limit:60 °C
- Kulstofbelastning:9%
- pH-område:2,0 - 9,0
- Surface area:160 m²/g