Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining racks with handle for 50 slides, - mm
Katalog # 631-2507
Leverandør:   DWK Life Sciences
Staining dishes, WHEATON®
Katalog # 631-2507
Leverandør:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining racks with handle for 50 slides, - mm
Price
2.860,00 kr
Log ind for tilgængelighed
1 * 3 Items
Product Details & Documents
Soda-lime glass and stainless steel.
  • Handle permanently attached
  • Hinged closure
  • Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Specifikationer
  • Beskrivelse:
    Staining racks with handle for 50 slides
  • B×D×H:
    - mm