Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
29/32, 60 mm
Katalog # COWI010.1229
Leverandør:   COWIE
Stoppers, self realasing, PTFE
Katalog # COWI010.1229
Leverandør:  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
29/32, 60 mm
Price
345,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifikationer
  • NS:
    29/32
  • Højde:
    60 mm