Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
pH-elektroder, InLab®

pH-elektroder, InLab®

InLab® Routine PT1000, InLab® Routine PT1000**, 0 - 14, 0...+100 °C, Glas, 3 mol/l KCl, Ceramic, 12×120 mm, MultiPin™, Drinking water, highly saline solutions, sea water, TRIS buffers, corrosive acids/bases
Katalog # 662-2832
Leverandør:   Mettler - Toledo
pH-elektroder, InLab®
Katalog # 662-2832
Leverandør:  Mettler - Toledo
pH-elektroder, InLab®
pH-elektroder, InLab®

InLab® Routine PT1000, InLab® Routine PT1000**, 0 - 14, 0...+100 °C, Glas, 3 mol/l KCl, Ceramic, 12×120 mm, MultiPin™, Drinking water, highly saline solutions, sea water, TRIS buffers, corrosive acids/bases

Price
3.210,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Med S7, BNC eller MultiPin™ (pH/temperatur) stik.

  • Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
  • ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
  • InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions

Junction: Movable glass sleeve

* integreret temperaturprobe (NTC30)
** integreret temperaturprobe (Pt1000)

Electrolyte: 3 mol/l KCl

Bestillingsoplysninger: Supplied with test certificate and with (BNC connection) or without cable (S7 and MultiPin™ connections). Junction tool and set of junctions for replacing the junction of the InLab® Versatile Pro electrode are available separately.

Specifikationer
  • Tilslutning:
    MultiPin™
  • Ø×L:
    12×120 mm
  • Til:
    Drinking water, highly saline solutions, sea water, TRIS buffers, corrosive acids/bases
  • Model:
    InLab® Routine PT1000**
  • Beskrivelse:
    InLab® Routine PT1000
  • Materiale:
    Glas
  • Seq:
    03
  • Membran:
    Ceramic
  • Elektrolyt:
    3 mol/l KCl
  • Temperatur:
    0...+100 °C
  • pH:
    0 - 14
Accessories

Produktinformation

  • Katalog #
    Længde
    Tilgængelighed
    Price
  • InLab® cable MultiPin-BNC/RCA
    InLab® cable MultiPin-BNC/RCA
    661-0249
    1,2 m
    Log ind for tilgængelighed
    1 * 1 Items - 1.230,00 kr

Produktinformation

  • Katalog #
    Længde
    Tilgængelighed
    Price
  • InLab® cable S7-BNC
    InLab® cable S7-BNC
    661-0257
    1,2 m
    Log ind for tilgængelighed
    1 * 1 Items - 564,00 kr

Produktinformation

  • Katalog #
    Længde
    Tilgængelighed
    Price
  • InLab® cable S7-DIN
    InLab® cable S7-DIN
    661-0261
    1,2 m
    Log ind for tilgængelighed
    1 * 1 Items - 630,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Set of 10 replaceable junctions
    Set of 10 replaceable junctions
    METT51340268
    Log ind for tilgængelighed
    1 * 1 Items - 507,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Junction tool, for exchanging the replaceable junction
    Junction tool, for exchanging the replaceable junction
    METT51340267
    Log ind for tilgængelighed
    1 * 1 Items - 670,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.