pH-elektroder, InLab®
InLab® Routine Pro, InLab® Routine Pro*, 0 - 14, 0...+100 °C, Glas, 3 mol/l KCl, Ceramic, 12×120 mm, MultiPin™, Drinking water, highly saline solutions, sea water, TRIS buffers, corrosive acids/bases
Med S7, BNC eller MultiPin™ (pH/temperatur) stik.
- Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
- ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
- InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions
Junction: Movable glass sleeve
* integreret temperaturprobe (NTC30)
** integreret temperaturprobe (Pt1000)
Electrolyte: 3 mol/l KCl
: Supplied with test certificate and with (BNC connection) or without cable (S7 and MultiPin™ connections). Junction tool and set of junctions for replacing the junction of the InLab® Versatile Pro electrode are available separately.
- Tilslutning:MultiPin™
- Ø×L:12×120 mm
- Til:Drinking water, highly saline solutions, sea water, TRIS buffers, corrosive acids/bases
- Model:InLab® Routine Pro*
- Beskrivelse:InLab® Routine Pro
- Materiale:Glas
- Seq:02
- Membran:Ceramic
- Elektrolyt:3 mol/l KCl
- Temperatur:0...+100 °C
- pH:0 - 14
Accessories
- BeskrivelseLængde
- BeskrivelseLængde
- BeskrivelseLængde
- Beskrivelse
- Beskrivelse



