pH-elektroder, InLab®
InLab® Science, InLab® Science, 0 - 12, 0...+100 °C, Glas, 3 mol/l KCl, Movable glass sleeve, 12×120 mm, S7, Wastewater, TRIS buffers, yeast fermentation solution, starch solution, organic solvents
Katalog # 662-2837
Leverandør: Mettler - Toledo
Gå til:
Product Details & Documents
Med S7, BNC eller MultiPin™ (pH/temperatur) stik.
- Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
- ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
- InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions
Junction: Movable glass sleeve
* integreret temperaturprobe (NTC30)
** integreret temperaturprobe (Pt1000)
Electrolyte: 3 mol/l KCl
Bestillingsoplysninger: Supplied with test certificate and with (BNC connection) or without cable (S7 and MultiPin™ connections). Junction tool and set of junctions for replacing the junction of the InLab® Versatile Pro electrode are available separately.
Documents
Certifikater
Søg efter partinummerMETT51343056_TDS_EN
Datablad
Mettler InLab® Sensors
Produktlitteratur
Specifikationer
- Tilslutning:S7
- Ø×L:12×120 mm
- Til:Wastewater, TRIS buffers, yeast fermentation solution, starch solution, organic solvents
- Model:InLab® Science
- Beskrivelse:InLab® Science
- Materiale:Glas
- Seq:05
- Membran:Movable glass sleeve
- Elektrolyt:3 mol/l KCl
- Temperatur:0...+100 °C
- pH:0 - 12
Accessories
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- InLab® cable MultiPin-BNC/RCA
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- InLab® cable S7-BNC
Produktinformation
- Katalog #LængdeTilgængelighedPrice
- InLab® cable S7-DIN
Produktinformation
- Katalog #TilgængelighedPrice
- Set of 10 replaceable junctions
Produktinformation
- Katalog #TilgængelighedPrice
- Junction tool, for exchanging the replaceable junctionJunction tool, for exchanging the replaceable junctionMETT51340267Log ind for tilgængelighed1 * 1 Items - 670,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



