Orders LinkContactUsLinkComponent
Stabilising rings for erlenmeyer
Stabilising rings for erlenmeyer
Stabilising rings, Erlenmeyer, 70 mm, 340
Katalog # BOCH5661
Leverandør:   BOCHEM
Stabilising rings for erlenmeyer
Katalog # BOCH5661
Leverandør:  BOCHEM
Stabilising rings for erlenmeyer
Stabilising rings for erlenmeyer
Stabilising rings, Erlenmeyer, 70 mm, 340
Price
370,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PVC covered steel rings.
Specifikationer
  • Vægt:
    340
  • Beskrivelse:
    Stabilising rings
  • Seq:
    02
  • Til:
    Erlenmeyer
  • Ø indv.:
    70 mm