Automatic burettes, Dr Schilling pattern, Schellbach stripe
50 ml, 0,1 ml, 0,100 ± ml
Katalog # HECH40820050
Leverandør: GLASWARENFABRIK KARL HECHT
Automatic burettes, Dr Schilling pattern, Schellbach stripe
Katalog # HECH40820050
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
AR-GLAS®, class B.
- Burettes are calibrated to deliver (marked 'Ex')
- Schellbach stripe and fused graduation, faceted outlet tip
- Improved special pinch cock with titration screw for precise regulation
Precisely calibrated, tolerance 1,5 times the official tolerance of class AS
Specifikationer
- Kapacitet:50 ml
- Seq:05
- Inddeling:0,1 ml
- Tolerance:0,100 ± ml
Accessories
Produktinformation
- Katalog #KapacitetInddelingTilgængelighedPrice
- Ekstra burette, AR®/sodaglasEkstra burette, AR®/sodaglasHECH408210055 ml0,05 mlLog ind for tilgængelighed1 * 1 Items - 400,00 kr
Produktinformation
- Katalog #KapacitetInddelingTilgængelighedPrice
- Ekstra plastikbase, størrelse 1, til 500 ml Schilling-flaskerEkstra plastikbase, størrelse 1, til 500 ml Schilling-flaskerHECH40823001--Log ind for tilgængelighed1 * 1 Items - 47,80 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



