Ethidiumbromid
Glass bottle for solids, 1 g
Katalog # RESERES3011E-A101X
Leverandør: RESEARCH ORGANICS
Gå til:
Product Details & Documents
Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
- CAS nummer:1239-45-8
- Formel:C₂₁H₂₀BrN₃
- MDL nummer:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:13,04751
- Kogende Pt:72 °C (0,1 mmHg)
- Smeltende Pt:261…264 °C
- Tæthed:1,01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:214-984-6
- FN:2811
- ADR:6.1,I
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g