Orders LinkContactUsLinkComponent
Temperature stabilisation jacket
Temperature stabilisation jacket
Quick-release closure
Katalog # SCOI285405549
Leverandør:   SI Analytics
Temperature stabilisation jacket
Katalog # SCOI285405549
Leverandør:  SI Analytics
Temperature stabilisation jacket
Temperature stabilisation jacket
Quick-release closure
Price
920,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
  • Support plate to facilitate the use
  • Quick-action seal to simplify the changing viscometers
  • Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.
Specifikationer
  • Beskrivelse:
    Quick-release closure
  • Seq:
    103