Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for LED cold light sources

Accessories for LED cold light sources

Objective adapter RL Øi = 66 mm to objective Ø = 60 mm
Katalog # 631-0894
Accessories for LED cold light sources
Katalog # 631-0894
Accessories for LED cold light sources
Accessories for LED cold light sources

Objective adapter RL Øi = 66 mm to objective Ø = 60 mm

Price
277,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Documents

Specifikationer
  • Beskrivelse:
    Objective adapter RL Øi = 66 mm to objective Ø = 60 mm
  • Seq:
    960