Nutrient Pad Sets, NKS e.motion
Tergitol TTC, 0,45 µm, Total count, 100
Katalog # SART14055-50-RDN
Leverandør: Sartorius
Gå til:
Product Details & Documents
Nutrient pad sets for colony counting by the membrane filter method, specially designed for the Microsart e.motion dispenser. These nutrient pad consist of dehydrated culture media inserted into pre-sterilised Petri dishes. This alternative to agar media has a long storage life of 24 months at room temperature.
- Economical, time-saving microbiological control
- High quality and reproducible results for up to 24 months
- Compliant with international standards and references
- Easy handling
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:100
- Porestørrelse:0,45 µm
- Model:Tergitol TTC
- Til:Total count