Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumsulfat ≥98%, TECHNICAL

Natriumsulfat ≥98%, TECHNICAL

Plastic bucket, 25 kg
Katalog # 28104.460
Leverandør:   VWR Chemicals
Natriumsulfat ≥98%, TECHNICAL
Katalog # 28104.460
Leverandør:  VWR Chemicals
CAS nummer:  7757-82-6
Natriumsulfat ≥98%, TECHNICAL
Natriumsulfat ≥98%, TECHNICAL

Plastic bucket, 25 kg

CAS nummer:  7757-82-6
Price
3.320,00 kr
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
  • CAS nummer:7757-82-6
  • Formel:Na₂SO₄
  • MDL nummer:MFCD00003504
  • Molecular Weight:142.04 g/mol
  • Merck Index:12,08829
  • Kogende Pt:1429 °C (1013 hPa)
  • Smeltende Pt:888 °C
  • Tæthed:2,7 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-820-9
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 98 %

Specifikationer

  • Pakning:
    Plastic bucket
  • Pakn:
    25 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.