Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Petriskåle

Petriskåle

Petri-skål, med låg, 75 mm, 20 mm, Ikke steril
Catalog # BOCH8636
Supplier:   BOCHEM
Petriskåle
Catalog # BOCH8636
Supplier:  BOCHEM
Petriskåle
Petriskåle

Petri-skål, med låg, 75 mm, 20 mm, Ikke steril

Price
169,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Ø:
    75 mm
  • Sterilitet:
    Ikke steril
  • Beskrivelse:
    Petri-skål, med låg
  • Højde:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.