Petriskåle
Petri-skål, med låg, 120 mm, 20 mm, Ikke steril
Catalog # BOCH8638
Supplier: BOCHEM
Jump to:
Product Details & Documents
Specifications
- Ø:120 mm
- Sterilitet:Ikke steril
- Beskrivelse:Petri-skål, med låg
- Højde:20 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



