Natriumsilikat in aqueous solution, TECHNICAL
Plastic drum, 25 L
Supplier: VWR Chemicals
Natriumsilikat in aqueous solution, TECHNICAL
Catalog # 28079.460
Supplier: VWR Chemicals
CAS Number: 1344-09-8
Jump to:
Product Details & Documents
Synonyms: Vandglas
- CAS Number:1344-09-8
- Formula:Na₂O(SiO₂)x·xH₂O
- MDL Number:MFCD00003492
- Density:1,34 g/cm³
- EINECS:215-687-4
- UN:3266
- ADR:8,III
Documents
Certificates
Search by Lot Number2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC28079.320_SPEC_EN.pdf P20_VWRC28079.363_SPEC_EN.pdf P20_VWRC28079.460_SPEC_EN.pdf spec_VWRC28079.320 spec_VWRC28079.320
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Silica:25.6 - 27.6 %
- Sodium oxide:7.5 - 8.5 %
Specifications
- Pakning:Plastic drum
- Pakn:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



