: Ethylmethylketon, Butanon
- :78-93-3
- :C₂H₅COCH₃
- :MFCD00011648
- :72.11 g/mol
- :13,06097
- :79,6 °C (1013 hPa)
- :–86 °C
- :0,8054 g/cm³ (20 °C)
- :–4 °C (DIN 51755 Part 1)
- :Stue temperatur
- :201-159-0
- :1193
- :3,II
- :01-2119457290-43
- Assay:Min. 99 %
- Boiling point:79 - 80 °C
- Density (20/4):0.804 - 0.807
- Pakning:Plastic bottle
- Pakn:2,5 L
- Cat. No.:25654.320
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



