Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 1 mm
Katalog # 710-1033
Leverandør: INTEGRA Biosciences
Gå til:
Product Details & Documents
Documents
Dose It
Videos
Specifikationer
- Beskrivelse:Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm
- Ø indv.:1 mm