Inserts for vials, WHEATON®
Limited volume insert for 1,8 ml vial, 0,25 ml, Flat bottom, Glas, No
WHEA225350-531
- Kasket inkluderet:No
- Kapacitet:0,25 ml
- Beskrivelse:Limited volume insert for 1,8 ml vial
- Materiale:Glas
- Grundlag:Flat bottom