Accessories
Closed base plate 900×520 mm, with soap and removable drip tray and mounting panel, Glassware washing units
Katalog # 141-0647
Gå til:
Discontinued
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifikationer
- Beskrivelse:Closed base plate 900×520 mm, with soap and removable drip tray and mounting panel
- Seq:06
- Til:Glassware washing units