Orders LinkContactUsLinkComponent
Accessories for titrators
Silicon tubing, 450 mm, for DL31 Karl Fischer titrator
Catalog # METT51107490
Supplier:   Mettler - Toledo
Accessories for titrators
Catalog # METT51107490
Supplier:  Mettler - Toledo
Accessories for titrators
Accessories for titrators
Silicon tubing, 450 mm, for DL31 Karl Fischer titrator
Price
176,00 kr
Log in for availability
1 * 1 Items
Specifications
  • Beskrivelse:
    Silicon tubing, 450 mm, for DL31 Karl Fischer titrator
  • Seq:
    46