Orders LinkContactUsLinkComponent
Universal couplings, BOLA
outer thread M 6, male, UNF ¼" 28 G, outer thread, PTFE
Katalog # BOHLF772-08
Leverandør:   Bohlender
Universal couplings, BOLA
Katalog # BOHLF772-08
Leverandør:  Bohlender
Universal couplings, BOLA
Universal couplings, BOLA
outer thread M 6, male, UNF ¼" 28 G, outer thread, PTFE
Price
535,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PTFE, white.
  • Temperature resistance: from –200 to +250 °C
  • Universal chemical resistance
  • FDA conform
Coupling for transition from M6 thread to UNF ¼" 28 G thread. The M6 thread is marked by a groove.
Specifikationer
  • Beskrivelse:
    outer thread M 6
  • Materiale:
    PTFE
  • Gevind:
    male, UNF ¼" 28 G, outer thread