Orders LinkContactUsLinkComponent
Lids for 96-well microplates
96-well microplate lid, high profile (9 mm), klar, Ikke steril
Katalog # 736-6550
Leverandør:   Greiner Bio-One
Lids for 96-well microplates
Katalog # 736-6550
Leverandør:  Greiner Bio-One
Lids for 96-well microplates
Lids for 96-well microplates
96-well microplate lid, high profile (9 mm), klar, Ikke steril
Price
657,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
These high profile lids, height 9 mm, are designed to protect against contamination and evaporation during sample storage and cell culture. Lids without condensation rings are mainly used with non tissue culture treated microplates. Lids with condensation rings are recommended for cell culture applications, since they inhibit cross contamination.
Specifikationer
  • Sterilitet:
    Ikke steril
  • Farve:
    klar
  • Beskrivelse:
    96-well microplate lid, high profile (9 mm)