Orders LinkContactUsLinkComponent
DNA ladder
DNA ladder
20 bp ladder, 25 fragments, with 200 ng/µl concentration, 100 Tests
Katalog # LONZ50330
Leverandør:   LONZA
DNA ladder
Katalog # LONZ50330
Leverandør:  LONZA
CAS nummer:  
DNA ladder
DNA ladder
20 bp ladder, 25 fragments, with 200 ng/µl concentration, 100 Tests
CAS nummer:  
Price
kr2,640.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
Specifikationer
Specifikationer
  • Pakn:
    100 Tests
  • Beskrivelse:
    20 bp ladder, 25 fragments, with 200 ng/µl concentration