DNA ladder
100 bp extended range ladder, 30 fragments, with 200 ng/µl concentration, 100 Tests
Katalog # LONZ50322
Leverandør: LONZA
Gå til:
Product Details & Documents
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
Specifikationer
Specifikationer
- Pakn:100 Tests
- Beskrivelse:100 bp extended range ladder, 30 fragments, with 200 ng/µl concentration