Propper til standardslib, glas
45/40, Brunt glas
Catalog # HIRS8902393
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Stoppers from clear or amber glass.
Certifications: DIN 12252
Specifications
- NS:45/40
- Model:Brunt glas
Stoppers from clear or amber glass.
Certifications: DIN 12252