Orders LinkContactUsLinkComponent
Tilbehør og reservedele
Membrane wrench for diaphragm change SW 4 (for ME 1, MD 1)
Catalog # VACU20637777
Supplier:   VACUUBRAND
Tilbehør og reservedele
Catalog # VACU20637777
Supplier:  VACUUBRAND
Tilbehør og reservedele
Membrane wrench for diaphragm change SW 4 (for ME 1, MD 1)
Price
107,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.
Specifications
  • Beskrivelse:
    Membrane wrench for diaphragm change SW 4 (for ME 1, MD 1)