Orders LinkContactUsLinkComponent
Agarose, SeaKem® LE
Agarose, SeaKem® LE
Agarose, SeaKem® LE
Katalog # 733-1537
Leverandør:   LONZA
Agarose, SeaKem® LE
Katalog # 733-1537
Leverandør:  LONZA
Agarose, SeaKem® LE
Agarose, SeaKem® LE
Agarose, SeaKem® LE
Price
3.320,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
SeaKem® LE is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
  • Quality tested to certify performance
  • High gel strength
  • Low EEO
  • Standard melting temperature
  • DNase- and RNase-free
SeaKem® LE agarose is ideal for routine analysis of nucleic acids by gel electrophoresis and blotting. Each SeaKem LE gel sharply resolves DNA and provides consistent resolution from lot-to-lot. This molecular biology grade agarose has no detectable DNase or RNase activity and forms strong gels with low background upon ethidium bromide, SYBR® Green, or GelStar™ staining. SeaKem® LE is also recommended for protein gel electrophoresis in applications such as Ouchterlony and radial immunodiffusion (RID).
Specifikationer
  • Beskrivelse:
    Agarose, SeaKem® LE
  • Seq:
    02