Automation tips for Tecan®, Axygen®
Product Details & Documents
PP, med eller uden filter, sterile eller ikke sterile.
- Black tips are conductive for liquid level-sensing protocols
- Optimised fine point for accurate dispensing
- Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Emballering: Packaged in plastic blister packs.
Advarsel: * Economy packed in cardboard wrapper.
Specifikationer
- Pakket:16 racks, 96 i hvert
- Steril:+
- Volumen:1000 µl
- Seq:08
- Spidstype:TT-1000-C-HTR-S
- Spits farge:Transparent
- Til brug med:LiHA-1, -4, -8
- Filter:-
- Cat. No.:732-0962
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning